Learn More
Wilmad Labglass TRAP VAC 2PC 48X250MM COMP
Supplier: Wilmad Labglass LG11026106
Vacuum Trap, Two-Piece, with Hose Connectors, Complete. Body O.D.: 48mm; Body Length: 250mm; Joint: [ST] 45/50; Hose Connectors: 3/8 in.. For removal of vapors from a vacuum line by condensation with liquid air or dry ice slurry mixture