Learn More
Wilmad Labglass TRAP VAC 2PC 45X250MM COMP
Supplier:  Wilmad Labglass LG11026104
Encompass_Preferred
Vacuum Trap, Two-Piece, with Hose Connectors, Complete. Body O.D.: 45mm; Body Length: 250mm; Joint: [ST] 40/50; Hose Connectors: 3/8 in.. For removal of vapors from a vacuum line by condensation with liquid air or dry ice slurry mixture
Catalog No. 50-806-715