Learn More
Ace Glass, Inc. Trap, bump, 100mL, 29/42 top, 14/20 bottom
SDP
Supplier:  Ace Glass, Inc. 670412
Encompass_Preferred
100mL bump trap for use between the rotary evaporators steam tube and evaporator flask, preventing fluid from being drawn into the condenser. Trap has 29/42 top joint and 14/20 bottom joint.
Catalog No. 50-973-366