Learn More
Cole-Parmer Cole-Parmer K1VAC/60PSI Sealed SS Compound Transducer, 30 "Hg-60 psi, 4-20 mA Out
Supplier: Cole-Parmer 0735613
Stainless steel diaphragm and polysilicon strain gauge are bonded by vapor deposition—no epoxies or glues which can cause signal instability or drift.