Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mirus Bio™ TransIT-VirusGEN™ Transfection Reagent
TransIT-VirusGEN™ Transfection Reagent is ideal for recombinant adeno-associated virus and lentivirus production.
Supplier: Mirus Bio™ MIR 6735
This item is not returnable.
View return policy
Description
TransIT-VirusGEN™ Transfection Reagent is designed to enhance delivery of packaging and transfer vectors to adherent and suspension HEK 293 cell types to increase recombinant adeno-associated virus (AAV) and lentivirus production.
- Reliable – Consistent high virus titer production
- Scalable – Efficient across different formats
- Flexible – Compatible with different recombinant virus and cell culture systems
- Animal Origin Free – High performance with maximum compatibility
Specifications
Store at -10°C to -30°C | |
293-derived Cells | |
None (Promoterless) | |
For preclinical and early phase clinical trials to enhance delivery of packaging and transfer vector DNA to suspension and adherent HEK 293 cell types, To increase production of recombinant lentivirus and AAV | |
Transfection Reagent | |
Sterile |
Before each use, warm to room temperature and vortex gently. | |
Bottle | |
Chemical Transfection | |
Bottle | |
150 mL |
Safety and Handling
ShelfLife : 6 months from date of purchase, when properly stored and handled.
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction