Learn More
Thomson Instrument Company Bidirectional Transfer Cap, 2' Tubing Weld to C-Flex 16, Sterile | Use w/ 931113 | CS8
Supplier: Thomson Instrument Company 9317048
Thomson Instrument Company Optimum Growth™ Bidirectional Transfer Caps (patented) with downstem, utilize a peristaltic pump for easy aseptic transfer of media or cells into or out of cell bags, bioreactors, and flasks (from any manufacturer). Specific Bidirectional Transfer Caps work with the 1.6L, 2.8L and 5L Thomson Optimum Growth™ Flasks and your peristaltic pump. Simply replace the vented cap included with each flask with the appropriate Transfer Cap and connect to your vessel of choice. Transfer tubing comes fitted with male Luer lock, or a plugged end ideal for tube fusing. These products are individually packaged and sterilized for immediate use. The Optimum Growth system feature a unique low shear baffle design which increases aeration while maintaining cell viability and improving yields. Thomson Instrument Company manufactures its products in the USA to assure same day shipping.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.