Learn More
Cole-Parmer Ashcroft T2100PSI Pressure Transmitter, DIN 43; 0 to 100 psi
Supplier: Cole-Parmer 6843961
With all-welded pressure construction, the transducer meets demanding requirements in general industrial applications. 0.25% accuracy class. Ranges 30 psi through 20,000 psi. Broad temperature capability. Proven polysilicon thin film sensor