Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TraceCERT™ Methyl parathion Solution, 1,000 μg/mL in Acetone, MilliporeSigma™ Supelco™
Supplier: MilliporeSigma Supelco 40572
Description
Agriculture and EnvironmentalSpecifications
Ampule | |
298-00-0 | |
TraceCERT | |
1,000 μg/mL in acetone |
1 mL | |
Certified Reference Material | |
TraceCERT™ Analytical Standard | |
-17°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC
Spot an opportunity for improvement?
Provide Content Correction