Learn More
Fisher Education™ Traceable™ Flip-Stick™ Refrigerator Thermometer, Timer, and Hygrometer Bundle
SCI_ED
Combines a Flip-Stick™ refrigerator thermometer, timer, and hygrometer
Supplier:  Fisher Education™ 0800052
Catalog No. S94143
Explore available promotions
Add to Cart
Description
Description
Traceable™ Flip-Stick™ refrigerator thermometer, timer, and hygrometer bundle.
  • Flip-Stick™ Refrigerator Thermometer part number 0800052
  • Timer part number 0800048
  • Hygrometer part number 0800047
Specifications
Specifications
Temperature, Humidity, Time
Flip-Stick™ Refrigerator Thermometer, Timer, and Hygrometer
± 1°C between –20 and 100°C
0.1° from –20 to 200°C; or 1°C elsewhere
-50°C to +300°C
0.0 to 90.0% RH
-58 to 572°F
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov