Learn More
Fisher Education™ Traceable™ Digital-Bottle™ Refrigerator Thermometer, Timer, and Hygrometer Bundle
SCI_ED
Combines a Digital-Bottle™ refrigerator thermometer, timer, and hygrometer
Supplier:  Fisher Education™ 0800049
Catalog No. S94142
Explore available promotions
Add to Cart
Description
Description
Traceable™ Digital-Bottle™ refrigerator thermometer, timer, and hygrometer bundle.
  • Digital-Bottle™ Refrigerator Thermometer part number 4426
  • Timer part number 0800048
  • Hygrometer part number 0800047
Specifications
Specifications
Temperature, Humidity, Time
Traceable™ Digital-Bottle™ Refrigerator Thermometer, Timer, and Hygrometer
±0.4°C 0.5°C between 0° to 50°C; otherwise ± 1°C
0.1°
-20°C to +50°C
0.0 to 90.0% RH
-4 to 122°F
Bottle probe
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov