Learn More
Thermo Scientific iConnect™ Flame Ionization Detector (FID) for TRACE™ 1300 and 1600 Series GC
Supplier: Thermo Scientific 19070001
Achieve extremely sensitive detection of organic compounds over a wide linear dynamic range using the Thermo Scientific™ iConnect™ Flame Ionization Detector (FID) for TRACE™ 1300 and 1600 Series GC systems. The detector's small cell volume and rapid acquisition response make it ideal for both standard and fast chromatography.
This item is not returnable.
View return policy