Learn More
Regis Technologies Inc GC Regeant, Regis, TPC (5 mL)
Supplier: Regis Technologies Inc 1440002200
TPC is an Acylation Reagent. TPC is the reagent of choice for the resolution of optically active amines by gas chromatography. TPC provides sample volatility and couples with amines to form diastereomers which can be separated on GC columns. Its use in the determination of amphetamines and other drugs of abuse testing has attracted considerable interest.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.