Learn More
Creative Biomart Recombinant T. gondii P32 Antigen, His-tagged
SDP
Supplier:  Creative Biomart TOX457
Encompass_Preferred
Recombinant Toxoplasma Gondii P32 (GRA6) produced in E. coli containing 180 amino acids of the Dense Granule Antigen 6 (GRA6) and fused with a 6 His tag at C-terminus. This antigen forms dimers on
Catalog No. 50-208-4835
Due to product restrictions, please to purchase or view availability for this product.