Learn More
Creative Biomart Recombinant T. gondii P22 Antigen, His-tagged
Supplier: Creative Biomart TOX456
Recombinant Toxoplasma Gondii P22 (SAG2) produced in E. coli is a full length Surface antigen 2 (SAG2) and fused with a 6 His tag at C-terminus.