Learn More
Genscript Corporation ToxinEraser™ Endotoxin Removal Resin
Supplier: Genscript Corporation L00402
The ToxinEraser™ Endotoxin Removal Resin utilizes immobilized polymixin B to bind and remove pyrogens from solution. The polymixins are a family of antibiotics that contain a cationic cyclopeptide with a fatty acid chain. Polymixin B neutralizes the biological activity of endotoxins by binding to the lipid A portion of bacterial lipopolysaccharide.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.