Learn More
Cosmo Bio Usa Inc TOUCH BURNER APT-3
Supplier: Cosmo Bio Usa Inc PHDAPT3EX
Touch Burner APT-3 is portable, multipurpose butane burner. It features reliable ignition from a battery-powered igniter, an internal storage compartment that can be charged rapidly with widely available inexpensive butane gas, continuously variable flame intensity. It can be set to operate continuously for two handed techniques (such as micropipette formation) or transiently with an easy one-handed touch.Feature and AdvantagesCompact, small footprint design fits easily on crowded lab benchesComfortable one-handed on/off operationPermits remaining gas amount to be easily monitoredAdjustable flame permits various applicationsDesigned to prevent wax accumulation in tubesEasy to refill with standard butane gas fuelFoolproof safety lock prevents accidental ignitionEngineered for highest user safety
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.