Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diazyme Laboratories Total Bile Acids Test Kit
Enzyme cycling half kit ready for both manual methods
Supplier: Diazyme Laboratories DZ042AK01
Description
- Amplifies signal for accurate and fast measurement using small sample up to 50% less than conventional NVT methods
- Intra-assay precision CV%: < 4%
- Inter-assay precision CV%: < 3%
- For in vitro quantitative determination of total bile acids in human serum
Specifications
510(k) cleared, Health Canada registered | |
R1: 2 x 30 mL, R2: 2 x 10 mL | |
Liquid |
Total Biles Acid Test Kit | |
0 to 180 μM |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction