Learn More
Tosoh Bioscience AIA-PACK™ Test Cups - TU (T4 Uptake)
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
Specifications
For Use With (Application) Test cup technology provides consistent and accurate assay results since all assays run the same way, every time
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Type Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Type Price Quantity & Availability  
21-279-145
View Documents
Tosoh Bioscience
025270
5 trays of 20 cups TU Test Cup
Each for $685.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
21-279-51
View Documents
Tosoh Bioscience
020370
12 x 1 mL TU Calibrator Set
Each for $145.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • These tiny, bar coded test cups contain pre-measured, single unit-dosed, dry reagents that are required for completing specific assays on Tosoh AIA™ systems
  • Saves hands-on time for operators and reduces waste
  • No pre-mixing or on-board refrigeration
  • Calibration stability lasts up to 90 days on most assays
Specifications
Specifications
Test cup technology provides consistent and accurate assay results since all assays run the same way, every time