Learn More
Simco Top Gun Ionizing Air Gun, for Electronics and Cleanroom Applications
Supplier: Simco 400510501
High-performance ionizing air gun designed for electronics and cleanroom applications. Balanced to 0+/-15V, Top Gun provides a high blow-off force for efficient cleaning and rapid static decay; operates on compressed air or nitrogen. A filter at the exit of the gun is 99.9% efficient at 0.1um. Ionization and airflow are electricallly activated. Electronically balanced ion output protects ESD-sensitive components and assemblies. Shockless ionizing electrode is located outside the air stream. Made of lightweight static-dissipative plastic, gun body weighs only 6.5 oz. (185g) and features a light-touch trigger switch. All functionality is built into the gun, including flow control valve, balance adjustment for calabration, and a two-level LED which indicates power and ionization. Made of lightweight static-dissipative plastic, gun body weighs only 6.5 oz. (185g) and features a light-touch trigger switch. All functionality is built into the gun, including flow control valve, balance adjust
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.