Learn More
Electron Microscopy Sciences K-kits Tool Box, Silver, 0.2 µm Gap
Supplier: Electron Microscopy Sciences K7261S
The full tool set includes K-kit holder, sample Loading Stage, Needle Pen, Gluing Stand, Channel Opener, Sealing Glue, Mounting Glue, Glass Slides, 6/pk of K-kits, Shipping Box (empty), and some replacement parts.
Gap: 0.2 µm