Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Tolvaptan
Potent and selective vasopressin V2 receptor antagonist; renoprotective and orally active
$267.96 - $1089.24
Specifications
Purity | >98% |
---|---|
Gene Alias | OPC-41061 |
Product Type | Tolvaptan |
Molecular Weight (g/mol) | 448.94 |
Inhibitors | Tolvaptan |
Description
Potent and selective competitive vasopressin V2 receptor antagonist (Ki values are 0.06 and 12.3 nM for V2 and V1a receptors respectively). Decreases urine osmolality and increases serum sodium concentrations. Delays the onset of end-stage renal disease in a mouse model of polycystic kidney disease. Exhibits myocardial and renal protective effects in hypertensive heart failure rats. Orally active.Specifications
>98% | |
Tolvaptan | |
Tolvaptan |
OPC-41061 | |
448.94 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title