Learn More
MilliporeSigma™ TNFalpha ELISPOT Antibody Pair, Mouse
Protein Interaction Antibody Pairs
Supplier: MilliporeSigma™ ELI018M
Description
Includes:
- Mouse TNFα Capture Antibody, clone 6B8. Quantity: 500 μg.
- Mouse TNFα Detection Antibody, clone MP6-XT22. Quantity: 100 μg.
- Streptavidin Alkaline Phosphatase. Quantity: 55 μL.
Specifications
Monoclonal | |
Mouse | |
Primary | |
2-8°C for up to six months | |
Purified |
1.0mg/mL (Capture antibody), 0.5mg/mL (Biotinylated detection antibody) | |
96-well Plates | |
Mouse | |
6B8 (Capture), MP6-XT22 (Detection) |