Learn More
Invivogen Ultrapure Lipopolysaccharide from Escherichia coli 055:B5 (LPS-B5), 5mg
Supplier: Invivogen TLRLPB5LPS
Specificity: TLR4 agonist, Working concentration: TLR4 activity 100 pg - 1 μg/ml, LPS is the principal component of Gram-negative bacteria that activates the innate immune system.