Learn More
Teledyne ISCO, Inc TLC PLATES SILICA 200/CS
Supplier: Teledyne ISCO, Inc 692203400
Plate, TLC; Normal -phase; RediSep; TLC results transfer directly to column chromatography, expediting your method development; Produce more pure compound in less time; 5 x 10cm; 200/BX