Learn More
Miles Scientific 150A SILICA GEL HL 250UM 20X20CM (25 PLATES/BOX)
Supplier: Miles Scientific P76011
These are 150A silica gel plates. They not only ease handling and sample application but also permit the use of up to 80% water in the developing solvent without loss of adherence of the absorbent layer to the glass plate. Organic binder plates are recommended for all TLC applications except those that use vigorous charring for visualization.