Learn More
Chemglass Life Sciences TLC Developing Chamber Complete, Cylindrical, 34/15 Joint Size, Fits Plate 1in X 3
Supplier: Chemglass Life Sciences CG118101
TLC Developing Chamber Complete, Cylindrical, 34/15 Joint Size, Fits Plate 1in X 3. Cylindrical glass chamber for development of TLC plates from 25 x 75mm to 50 x 200mm. Chamber has a short length standard taper outer joint. Stopper is a short length inner with a tool ball style handle. The CG-1181-01 and -02 chambers have a hex base for added stability. Complete chamber is supplied with bottle and stopper.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.