Learn More
Chemglass Life Sciences 125mL TLC Chromatography Sprayer, Improved
Supplier: Chemglass Life Sciences CG118003
125mL TLC Chromatography Sprayer. Sprayer provides a fine, uniform spray ideally adjusted for the development of chromatographic patterns. Spray pattern is adjustable by varying the coverage of the vent hole with the users thumb. Sprayer is used with low pressure air (5/PSI) or rubber pressure bulb. Sprayer consists of an atomizer top attached to the reservoir with a Rodaviss 24/40 joint. This design provides for a positive, grease-free seal between the atomizer and reservoir and eliminates the glass hooks used on other sprayers. Complete sprayer consists of the atomizer top, Erlenmeyer shaped reservoir, CG-182-03 open-top screw cap and CG-305-212, #212 Viton o-ring.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.