Learn More
Agilent Technologies TKB1 COMPETENT CELLS
Supplier: Agilent Technologies 200134
TKB1 Competent Cells. Derived from BL21 strain, use with pET, pCAL or other T7 RNA promoter-based vectors. For producing tyrosine-phosphorylated proteins in vivo, Applications: Affinity screening of libraries, purification of proteins