Learn More
Chemglass Life Sciences 50mL Titrette Bottle Top Burette, Complete
SDP
Supplier:  Chemglass Life Sciences CG1161200
Encompass_Preferred
50mL Titrette Bottle Top Burette, Complete. The CG-1161 Titrette is the new generation of bottletop burette from BRAND. It has a number of innovative features and upgrades for more accurate and convenient titration. The Titrette avoids the risk of spills from poured transfers to glass or plastic burettes. The instrument if well suited for general chemistry, water treatment applications, food and beverage analysis, industrial titrations and environmental work. Titrette is supplied complete with the bottle top burette having a GL-45 Thread, telescoping filling tube 170-330mm in length, recirculation tube, 2 batteries, 2 colored light shield inspection windows, three bottle adapters: GL45 to 33-400 GPI thread, GL45 to S40 European Socket Adapter and GL45 to 29/32 Inner Joint, operating manual and 1 year warranty. For GL-45 bottles see CG-8088 or CG-8090.
Catalog No. 50-131-1000