Learn More
Chemglass Life Sciences 25mL Titrette Bottle Top Burette, Complete
SDP
Supplier:  Chemglass Life Sciences CG1161100
Encompass_Preferred
The CG-1161 Titrette is the new generation of bottle top burette from BRAND. It has a number of innovative features and upgrades for more accurate and convenient titration. The Titrette avoids the risk of spills from poured transfers to glass or plastic burettes. The instrument if well suited for general chemistry, water treatment applications, food and beverage analysis, industrial titrations and environmental work.
Catalog No. 50-131-0999