Learn More
Cole-Parmer Titrette 4760161 Digital Bottletop Burette/Dispenser; 50 mL
Supplier: Cole-Parmer 0791053
Burette (burets) switches automatically between filling and titrating modes. View dispensed results on the LCD readout with a resolution of 0.01uL. Unit can be easily and quickly disassembled for simple maintenance and cleaning, no servicing is required. Titrating tube can be adjusted horizontally and vertically. Suitable for aqueous or nonaqueous solutions up to 2N.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.