Learn More
Hach Company EDTA Digital Titrator Cartridge, 0.0716 M
Supplier: Hach Company 2081701
EDTA standard solution. For the determination of iron by Digital Titrator titration. 0.0716 M. Digits x 0.1 = mg/L Fe in 50 mL