Learn More
Hach Company Digital Titrator (w/o case)
Supplier: Hach Company 1690002
Portable and robust digital Titrator for fast and precise titrations. High resolution (125 µL/digit) and high accuracy (±1%). Easy handling.