Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Titration Solvent, MilliporeSigma™
Solvent for volumetric Karl Fischer titration with two component reagents Aquastar™.
$135.90 - $283.50
Specifications
Density | 0.90 g/cm3 (at 20°C) |
---|---|
Flash Point | 11°C (Methanol) |
Packaging | Glass Bottle |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
M1880151000
|
MilliporeSigma
1880151000 |
1 L |
Each for $135.90
|
|
|||||
M1880152500
|
MilliporeSigma
1880152500 |
2.5 L |
Each for $283.50
|
|
|||||
Specifications
0.90 g/cm3 (at 20°C) | |
11°C (Methanol) | |
Glass Bottle |
Safety and Handling
Recommended Storage : Store at 15 to 25°C
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title