Learn More
Andwin Scientific Tissue-Tek® Accu-Edge® Disposable Microtome Blades, Sakura® Finetek, High-Profile Blades, Pack of 50
SDP
Supplier:  Andwin Scientific 4685PK
Encompass_Preferred
The ultra-sharp Accu-Edge disposable high-profile blades by Sakura Finetek are compatible with all Accu-Edge blade holders that are specially designed for disposable blades 14 millimeters in height. This product is ideal for microtomy, cryotomy, and sectioning specimens without striations, distortion, or chattering. It is sold in dispensers that can store 50 blades and distribute them in a few seconds. The previously used blade is then automatically ejected.
Catalog No. NC9998535