Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Tissue Path™ Paraplast™ Tissue Embedding Media
Provides better tissue infiltration and superior quality sections than other embedding media
Catalog Number | Product Type | Melting Point | For Use With (Application) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Product Type | Melting Point | For Use With (Application) | Price | Quantity & Availability | ||||
23-021399
|
Tissue Embedding Media | 56°C | Provides better tissue infiltration and superior quality sections than other embedding media |
Item Discontinued
This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. View Alternate Product |
N/A | ||||
23-021401
|
Tissue Embedding Media | 50° to 54°C | Exceptional compression resistance and ribbon continuity |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View Alternate Product |
N/A | ||||
Description
- Greatly minimizes tissue compression due to sectioning
- Melting point of 122° to 129°F (50° to 54°C) allows lower infiltration temperatures
- Packaged in cases of eight, 1kg packs
- Paraplast PLUS is recommended for large tissues or tissue that normally presents infiltration problems and contains a small amount of DMSO (dimethyl sulfoxide) for faster tissue penetration
- Paraplast X-TRA is a unique blend of low molecular weight polymers and highly purified paraffins for exceptional compression resistance and ribbon continuity
Specifications
Tissue Embedding Media | |
8 Cases |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title