Learn More
Cole-Parmer Electrothermal Tissue float bath; 230VAC, 50/60Hz
Supplier: Cole-Parmer 4857255
This round bath unit has a temperature range from 86 to 158°F (30 to 70°C). Adjust temperature easily by simply turning the analog dial. The convenient edge serves as drying surface for processed slides. The black anodized bath surface reduces eye strain by cutting down on glare while improving visibility of tissue sections. Exterior is white powder-coated aluminum, making it easy to keep clean. Safety features include an amber neon light alerting you when heater is on, as well as automatic shut-off. Order gelatin separately to increase surface tension of water in flotation bath.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.