Learn More
Electron Microscopy Sciences Tissue Embedding Center
SDP
Supplier:  Electron Microscopy Sciences 62300
Encompass_Preferred
Request Bulk Order
The tissue embedding center has the latest in circuit board and component technology. Uniquely designed with microprocessor temperature sensors, a single console which maximizes space utilization, high-contrast viewing areas and multifunctional work stations. The entire unit is encased in a durable, non-heat conductive housing and is controlled by a microprocessor for dependability and performance.
Features
4-liter capacity paraffin reservoir which minimizes refilliling frequency.
Heated forceps warmer and illumniated paraffin dispenser.
Warming oven with removable shelf and double hinged lid
Wax bath complete with drainage shelf, debris screen, and hinged lid.
Catalog No. 50-293-24
Edge
Explore available promotions