Learn More
Tisch Scientific PM2.5 PTFE membrane filters are designed for PM2.5 air monitoring.
Supplier:  Tisch Scientific SF18040
Encompass_Preferred
Tisch PM2.5 PTFE membrane filters are designed for PM2.5 air monitoring. These high-purity, thin filtration devices are sequentially numbered and housed in chemically resistant polypropylene support rings for ambient air sampling.
Catalog No. 50-225-1426