Learn More
Labcon North America TipTop Dispenser Caps for Blood Collection Tubes, 16mm, in Bags
Supplier: Labcon North America 55128000009
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Dispenser; Blood Collection Tube; Tip Top™; Labcon; Dispenser cap with integral filter for 16mm blood collection tubes; Dispenses drops of filtered blood for clinical lab uses; Fits standard glass or plastic 16mm blood tubes; Polyethylene USP Classs VI resin; 1000 per bag/5 bags per case; Made in USA