Learn More
Tin plating powder, electroless, part A, Thermo Scientific Chemicals
$151.00
Specifications
Chemical Name or Material | Tin plating powder |
---|---|
Name Note | Electroless, part A |
MDL Number | MFCD03703330 |
UN Number | UN1759 |
Form | Powder |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA44176P2
|
Thermo Scientific Chemicals
044176P2 |
1 lb. |
Each of 1 for $151.00
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
To preserve solderability of printed circuit boards, for plating electronic parts, and for corrosion protection
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- To preserve solderability of printed circuit boards, for plating electronic parts, and for corrosion protection
Specifications
Tin plating powder | |
MFCD03703330 | |
Powder |
Electroless, part A | |
UN1759 |
Safety and Handling
GHS H Statement
H314-H351-H361-H302-H317
Causes severe skin burns and eye damage.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
May cause an allergic skin reaction.
P201-P202-P234-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P390-P501c
H290-H302-H314-H317-H335-H350-H361d-H373
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.