Learn More
Kirby Risk Electrical Supply Cable Tie Hand Tool | 10.4 Ounces
Supplier: Kirby Risk Electrical Supply GTSE
The GTS-E Ergonomic Cable Tie Hand Tool is easy to operate and has tool-controlled tension that provides a flush cut-off and speeds up installation to lower installed cost. It promotes worker safety, reduces downtime, and increases productivity. This tool is for subminiature, miniature, intermediate, and standard cross section cable ties. The tool weighs 10.4 ounces.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.