Learn More
CUSABIO BIOTECHNOLOGY LLC Rat Thyroid Stimulating Hormone,TSH ELISA Kit
Supplier: CUSABIO BIOTECHNOLOGY LLC CSBE05115R
This Rat TSH ELISA Kit was designed for the quantitative measurement of Rat TSH protein in serum, plasma, tissue homogenates, cell lysates. It is a Sandwich ELISA kit, its detection range is 0.6 μIU/mL-24 μIU/mL and the sensitivity is 0.3 μIU/mL. One kit contains 96 wells.