Learn More
Cliawaived Inc ThyroChek TSH Testing Kit (Whole Blood)
SDP
Supplier:  Cliawaived Inc THYROCHEK120AKIT
Encompass_Preferred
(20 Tests/Box) Controls NOT included.Thyroid (TSH) Hormone Testing Kits, FDA-cleared and CLIA waived thyroid test. ThyroChek TSH Tests check for elevated thyroid stimulating hormone (TSH) in whole blood. The ThyroChek test requires one drop of blood from a finger stick or venous draw. Results in 10 min. FDA Cleared and CLIA Waived. (FDA 510K#: K990658)
Catalog No. NC1859632
Due to product restrictions, please to purchase or view availability for this product.