Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Thrombo-Wellcotest™ Rapid Latex Agglutination Test
Rapidly detect fibrin and fibrinogen degradation products in human serum and urine by latex agglutination.
Catalog Number | Mfr. No. | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
R30852301
|
Thermo Scientific™
R30852301 |
Thrombo-Wellcotest Latex | 3 mL/Vial | 2°C to 8°C | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. | N/A | |||
R30852601
|
Thermo Scientific™
R30852601 |
Thrombo-Wellcotest Kit | 20 Tests/Kit | 2°C to 8°C | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. | N/A | |||
R30853001
|
Thermo Scientific™
R30853001 |
Thrombo-Wellcotest Collection Tubes | 100 Tubes/Pk. | 2°C to 8°C | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
R30853101
|
Thermo Scientific™
R30853101 |
Thrombo-Wellcotest Glycine Saline Buffer | 100 mL/Bottle | 2°C to 8°C | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
Description
Carry out semi-quantitative detection of fibrin and fibrinogen degradation products in human serum and urine using Thermo Scientific™ Thrombo-Wellcotest™ Rapid Latex Agglutination Test.
Use this simple, rapid, semi-quantitative test for the presence of fibrin/fibrinogen degredation products (FDP) in human serum and urine samples.
The test is designed as a slide agglutination method in which one drop of sample and one drop of latex suspension are mixed. A distinct agglutination pattern at the end of the test period indicates the presence of 2μg/ml FDP or more in the sample under test. By assaying samples at two different dilutions, an approximate FDP value can be determined.
Capable of detecting the presence of all the major breakdown products of fibrin or fibrinogen. The levels measured by this latex test correlate well with results obtained with a haemagglutination-inhibition immunoassay.
The following products are available for use with Thrombo-Wellcotest Test Kit and are sold separately:
The test is designed as a slide agglutination method in which one drop of sample and one drop of latex suspension are mixed. A distinct agglutination pattern at the end of the test period indicates the presence of 2μg/ml FDP or more in the sample under test. By assaying samples at two different dilutions, an approximate FDP value can be determined.
Capable of detecting the presence of all the major breakdown products of fibrin or fibrinogen. The levels measured by this latex test correlate well with results obtained with a haemagglutination-inhibition immunoassay.
The following products are available for use with Thrombo-Wellcotest Test Kit and are sold separately:
Thrombo-Wellcotest Latex (Part No. R30852301)
Thrombo-Wellcotest Collection Tubes (Part No. R30853001)—Glass tubes contain soya bean trypsin inhibitor (approximately 3600 NF units/tube) and Bothrops atrox venom (>10μg/tube) for the collection of 2mL whole blood or urine.
Thrombo-Wellcotest Glycine Saline Buffer (Part No. R30853101)—Glycine saline buffer is provided as a replacement item in instances where multiple tests have exhausted the supply of this component.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
2°C to 8°C |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title