Learn More
Thomas Scientific BLADES. PK 25 pk 05
Supplier: Thomas Scientific 6727C18
Thomas Tissue Slicer, Carbon Steel Blades, 25Ct; For preparation of thin, uniform sections of fresh tissue in metabolic studies of tissue in vitro; Sections average 0.5mm in thickness