Learn More
Bioworld THIOPHILIC SEPAROPOREAGAR
Supplier: Bioworld 201810382
THIOPHILIC SEPAROPORE (AGAROSE) 4B-CL: The term Thiophilic refers to an affinity for sulfone groups that lie in close proximity to thioether groups. Thiophilic affinity purification is based on the ability of some proteins, particularl...