Learn More
Cole-Parmer Digi-Sense Food Infrared (IR) Thermometer
Supplier:  Cole-Parmer 3562545
Encompass_Preferred
Temperature measurement is essential when ensuring proper food storage, cooking and serving environments, and is required to be FDA compliant. Quick checks with a non-contact thermometer can remove the hazard of cross-contamination and reduces cycle time over a traditional penetration probe. This thermometer provides this safe, non-contact reading in under a second. Use the TempTestr to monitor HACCP danger zone temperature standards for food (40 to 140degF or 4 to 60degC), the critical range where harmful bacteria grows most rapidly. LED’s on the meter quickly indicate if the food temperature is within a green or red bacterial growth zone. A green LED light indicates food-safe hot and cold holding temperatures. Red LED light indicates that food is exposed to potentially dange
Catalog No. 50-196-1555