Learn More
MilliporeSigma™ Supelco™ Thermogreen™ LB-1 Septa, Solid Disc
Suitable for conveniently introducing the sample without causing the system to leak. Improper septa decompose under the high-temperature conditions of capillary inlets causing baseline disruption and ghost peaks in a chromatogram.
Supplier: MilliporeSigma™ Supelco™ 20657U
Description
For use with:
- Minimizing extraction onto or loss of analytes off the solid phase microextraction (SPME) fiber from the vial during analysis by GC-MS
- Avoiding carrier gas leaks, extraneous peaks, and phthalate contamination in GC instruments
- Use in conjunction with a thick needle protector to avoid damage to GC instruments irreversibly caused by microsyringes
- Influent sampling ports in gas chromatographs
Specifications
13/32 in. | |
10 mm | |
50/Pk. | |
50°C to 300°C | |
Septa, LB-1 |
Green | |
Gas Chromatography | |
Solid Disc | |
Thermogreen |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Thermogreen is a registered trademark of Sigma-Aldrich Co. LLC