Learn More
Thermo Scientific™ ThermoFlex™ Recirculating Chillers
Optimized for the most demanding applications, high-performance ThermoFlex Recirculating Chillers are reliable, easy-to-use and easy-to-maintain.
Supplier: Thermo Scientific™ 121121010000001
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Description
Optimized for the most demanding applications, Thermo Scientific™ ThermoFlex Recirculating Chillers are reliable, easy-to-use and easy-to-maintain. The ThermoFlex platform was developed with customer input from concept to design. This high-performance chiller platform, with cooling capacity up to 24,000 watts (81,948 btu/hr), is configurable to meet application demands.
ThermoFlex Chillers are now available in low noise select models. Quiet performance (<58 dBA) promotes a safe and reduced stress environment in your lab. These low noise models are designed to remain quiet over the entire operating range of the chiller while maintaining temperature control accuracy.
Ease of Use
- An intuitive user interface for ease of operation
- Air and water filters that can be changed while unit is in operation
- Innovative packaging for rapid installation
- Quick start guide enables trouble-free system start-up
Superior Performance
- Improved cooling capacity
- Increased reliability
- Ease of maintenance
Configurable Design
- Wide range of available cooling capacities
- Variety of available options
- Installation flexibility
Recommended for:
- Aerospace
- Analytical
- Biotech
- Chemical
- Industrial
- Medical
- Pharmaceutical
- Research
- University
Specifications
10°C to 40°C | |
2500w; Pumping Option PD1; 208V 60Hz | |
NRTL Certified to CSA Standards; CE marked | |
5°C to 40°C | |
208 V 60 Hz | |
60 Hz |
1.9 gal. | |
7.2 L | |
2500 W | |
±0.1°C | |
208 V |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.